Magnetic stirring bars
Magnetic disc stirring bar Ø 10×20 mm, 20 mm, 10 mm, AREC series
VELPA00000355
- Ø:10 mm
- Til:AREC series
- Beskrivelse:Magnetic disc stirring bar Ø 10×20 mm
- Længde:20 mm
- Seq:09