[EN]CLOG ALTHEA WHITE OB E A SRC S.37 1 * 1 Par
Catalog # MILEALAB9/37
Supplier: MILLE S.A.S
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
VWR®, Serologiske pipetter, Standard type
Supplier: VWR Collection
Handsker, nitril, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR®, Ansigtsservietter
Supplier: VWR Collection
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
pH-teststrimler, MQuant®, Supelco®
Supplier: Merck
VWR®, Nitril handsker, NITRILE
Supplier: VWR Collection
Vand, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection