Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Redox standards, Supelco®

Redox standards, Supelco®

200 mV (25°C), ±5 mV, 500 mL
Katalog # 40661-500ML
Leverandør:   Merck
Redox standards, Supelco®
Katalog # 40661-500ML
Leverandør:  Merck
CAS nummer:  
Redox standards, Supelco®

200 mV (25°C), ±5 mV, 500 mL

CAS nummer:  
Price
935,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents

The values are the potential of a platinum electrode versus an Ag/AgCl reference in 3M potassium chloride.

Documents

Specifikationer

Specifikationer

  • Pakn:
    500 mL
  • Beskrivelse:
    200 mV (25°C)
  • Nøjagtighed:
    ±5 mV

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.