124 mV (25°C), ±5 mV, 500 mL
The values are the potential of a platinum electrode versus an Ag/AgCl reference in 3M potassium chloride.
#: 91233.270
: VWR Chemicals
#: 1.06657.1000
: Merck
#: 613-6774
: EPPENDORF
#: 1.10011.0002
#: 1.16977.0001
#: HACH1455232
: Hach
#: 631-0724
: VWR Collection
#: 1.70236.0100