Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Redox standards, Supelco®

Redox standards, Supelco®

650 mV (25°C), ±5 mV, 500 mL
Katalog # 87783-500ML
Leverandør:   Merck
Redox standards, Supelco®
Katalog # 87783-500ML
Leverandør:  Merck
CAS nummer:  
Redox standards, Supelco®

650 mV (25°C), ±5 mV, 500 mL

CAS nummer:  
Price
935,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents

The values are the potential of a platinum electrode versus an Ag/AgCl reference in 3M potassium chloride.

Documents

Specifikationer

Specifikationer

  • Pakn:
    500 mL
  • Beskrivelse:
    650 mV (25°C)
  • Nøjagtighed:
    ±5 mV

Specifikationer