Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Super Adjustable™ Super Erecta® shelving

Super Adjustable™ Super Erecta® shelving

Super Adjustable™ wire shelf, A3048NC, Forkromede, 1219×760×1219 mm, 9,5 kg
Katalog # METOA3048NC
Leverandør:   Metro International
Super Adjustable™ Super Erecta® shelving
Katalog # METOA3048NC
Leverandør:  Metro International
Super Adjustable™ Super Erecta® shelving
Super Adjustable™ Super Erecta® shelving

Super Adjustable™ wire shelf, A3048NC, Forkromede, 1219×760×1219 mm, 9,5 kg

Price
2.220,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Designed to work in conjunction with the entire Super Erecta® System of shelves and accessories, these Super Adjustable wire shelves have a unique corner release system which allows them to be adjusted quickly and easily without the use of tools.

  • Heavy duty construction
  • Built to save space and to stand though against years of heavy use
  • Adjust and add shelves quickly and easily to reclaimwasted space

They shelf have a sleek chrome finish that makes it perfect for dry storage areas, while the open-wire construction minimises dust accumulation and allows a free circulation of air and greater visibility.

Specifikationer
  • Beskrivelse:
    Super Adjustable™ wire shelf, A3048NC
  • Materiale:
    Forkromede
  • Seq:
    24
  • B×D×H:
    1219×760×1219 mm
  • Vægt:
    9,5 kg