96-Well plate sample control system, U-2D™
U-2D™ rack with 96 well plate, 1 ml preloaded inserts
Katalog # MICS95296-500-CS
Leverandør: MICROSOLV
Gå til:
Product Details & Documents
The U-2D™ universal two dimensional 96-well plate system offer users unprecedented sample management control compared to current glass lined or plastic well plates for sample preparation or chromatography.
Easily push up the insert from the bottom and remove it from the rack. These inserts can then be placed into any MicroSolv 9 mm screw top autosampler vial for safe storage, transport or for a second chromatography dimension, or to another instrument platform.
Ordering information: Base and sealing mats sold separately.
Specifikationer
- Beskrivelse:U-2D™ rack with 96 well plate, 1 ml preloaded inserts
- Seq:30
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Paddles for U-2D™ sealing film
Produktinformation
- Katalog #TilgængelighedPrice
- Roller for U-2D™ sealing film
Produktinformation
- Katalog #TilgængelighedPrice
- U-2D™ plate, 350µL, with rack and base onlyU-2D™ plate, 350µL, with rack and base onlyMICS95296-U2DLog ind for tilgængelighed1 * 1 Items - 865,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Zone free sealing film for U-2D™ platesZone free sealing film for U-2D™ platesMICS95296-ZFF-PKLog ind for tilgængelighed1 * 10 Items - 409,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Zone free sealing film for U-2D™ platesZone free sealing film for U-2D™ platesMICS95296-ZFF-CSLog ind for tilgængelighed1 * 50 Items - 1.710,00 kr