Gemfibrozil 98%
Glass bottle, 25 g
Katalog # ACRO461790250
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: CI-719
- CAS nummer:25812-30-0
- Formel:C₁₅H₂₂O₃
- MDL nummer:MFCD00079335
- Molecular Weight:250.34 g/mol
- Smeltende Pt:60…61 °C
- Opbevaringstemperatur:Fryser
Specifikationer
Specifikationstestresultater
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:58 to 63 °C
- Water:≤0.25 %
- HPLC:≥97.5 %
Specifikationer
- Pakning:Glass bottle
- Pakn:25 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



