Gemfibrozil 98%
Glass bottle, 5 g
Catalog # ACRO461790050
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: CI-719
- CAS Number:25812-30-0
- Formula:C₁₅H₂₂O₃
- MDL Number:MFCD00079335
- Molecular Weight:250.34 g/mol
- Melting Pt:60…61 °C
- Storage Temperature:Fryser
Specifications
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:58 to 63 °C
- Water:≤0.25 %
- HPLC:≥97.5 %
Specifications
- Pakning:Glass bottle
- Pakn:5 g
Specifications
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



