Orders LinkContactUsLinkComponent
Double spatulas, bent
Double spatulas, bent
185 mm, 10 mm
Catalog # USBE3293
Supplier:   USBECK Laborgeräte
Double spatulas, bent
Catalog # USBE3293
Double spatulas, bent
Double spatulas, bent
185 mm, 10 mm
Price
30,50 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Stainless steel.
Specifications
  • Længde:
    185 mm
  • Seq:
    02
  • Bladbredde:
    10 mm