Original Perfusor® syringes
Original Perfusor® syringe 50 ml, with particle filter cannula, type UV-Protect, orange-transparent, 50 ml, — ml, — ml, — mm, Centric cone
Katalog # BRAU8728861F-06
Leverandør: B. Braun
Gå til:
Product Details & Documents
PP syringe with safe plunger backstop and minimal residual volume.
- Oval grip plate for handling of the syringe and for prevention of rolling
- Plunger stopper with double sealing ring for slow aspiration or infusion of drug solutions, made of synthetic material or natural latex-free
- Centric Luer Lock cone for the attachment of a needle or infusion tubing
Documents
Specifikationer
- Udv.Ø:— mm
- Model:Centric cone
- Kapacitet:50 ml
- Beskrivelse:Original Perfusor® syringe 50 ml, with particle filter cannula, type UV-Protect, orange-transparent
- Længde:— ml
- Seq:07
- Inddeling:— ml