n-Hexan, anhydrous (max. 0.003% H₂O)
Glass bottle with septum cap, 500 mL
Leverandør: VWR Chemicals
n-Hexan, anhydrous (max. 0.003% H₂O)
Katalog # 85819.269
Leverandør: VWR Chemicals
CAS nummer: 110-54-3
Gå til:
Product Details & Documents
Synonymer: Hexyl hydride, Hexanen, Hexane
- CAS nummer:110-54-3
- Formel:H₃C(CH₂)₄CH₃
- MDL nummer:MFCD00009520
- Molecular Weight:86.18 g/mol
- Merck Index:13,04712
- Kogende Pt:69 °C (1013 hPa)
- Smeltende Pt:–94,3 °C
- Tæthed:0,659 g/cm³ (20 °C)
- Flash Pt:–22 °C
- Opbevaringstemperatur:Stue temperatur
- FN:1208
- ADR:3,II
- :0,2 µm Filtreret. Packaged under inert gas.
- EINECS:203-777-6
Specifikationer
Specifikationstestresultater
- Assay (GC, on anhydrous basis):99-100 %
- Color (APHA, Pt-Co):0-10
- Water (KF):0-0.003 % w/w
- Acidity (as Acetic acid):0-0.001 %
- Residue after evaporation:0-0.0005 % w/w
Specifikationer
- Pakning:Glass bottle with septum cap
- Pakn:500 mL



