Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Streptomycin sulfat

Streptomycin sulfat

Glass bottle, 100 g
Katalog # ACRO455341000
Irriterende
Sundhed
Leverandør:   Thermo Fisher Scientific
Streptomycin sulfat
Katalog # ACRO455341000
CAS nummer:  3810-74-0
Streptomycin sulfat
Streptomycin sulfat

Glass bottle, 100 g

CAS nummer:  3810-74-0
Price
1.090,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
  • CAS nummer:3810-74-0
  • Formel:C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
  • MDL nummer:MFCD00037023
  • Molecular Weight:1457.4 g/mol
  • Opbevaringstemperatur:Køleskab
Specifikationer

Specifikationstestresultater

  • Appearance:
    White to off-white Powder
  • Loss on drying:
    ≤7 %
  • Sulfate (SO4):
    18.0 to 21.5

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    100 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.