Iodmethan 99% stabilised
Glass bottle, 100 g
Katalog # ACRO459011000
Leverandør: Thermo Fisher Scientific
Iodmethan 99% stabilised
Katalog # ACRO459011000
Leverandør: Thermo Fisher Scientific
CAS nummer: 74-88-4
Gå til:
Product Details & Documents
Synonymer: Methyliodid
- CAS nummer:74-88-4
- Formel:CH₃I
- MDL nummer:MFCD00001073
- Molecular Weight:141.94 g/mol
- Kogende Pt:42,5 °C (760 mmHg)
- Smeltende Pt:–66 °C
- Tæthed:2,28 g/cm³
- Opbevaringstemperatur:Stue temperatur
- FN:2644
- ADR:6.1,I
Documents
Certifikater
Søg efter partinummerSpecifikationer
Specifikationstestresultater
- Appearance:Clear yellow to orange Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5290 to 1.5310 (20°C, 589 nm)
- GC:≥98.5 %
- Stabilizer:Copper
Specifikationer
- Pakning:Glass bottle
- Pakn:100 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



