Fenofibrate 98%
Glass bottle, 100 g
Katalog # ACRO461781000
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionic Acid Isopropyl Ester, Isopropyl 2-(4-(4-chlorobenzoyl)phenoxy)-2-methylpropanoate, Isopropyl 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionate
- CAS nummer:49562-28-9
- Formel:C₂₀H₂₁ClO₄
- MDL nummer:MFCD00133314
- Molecular Weight:360.83 g/mol
- Smeltende Pt:79…82 °C
- Opbevaringstemperatur:Stue temperatur
Documents
ACRO461781000_TDS_EN
Product specification
Specifikationer
Specifikationstestresultater
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:79 to 83 °C
- Loss on drying:≤0.5 % (60 °C) (vacuum)
- HPLC:≥97.5 %
Specifikationer
- Pakning:Glass bottle
- Pakn:100 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



