1,3-Propansulton 97%
Glass bottle, 1 kg
Leverandør: Thermo Fisher Scientific
1,3-Propansulton 97%
Katalog # ACRO131420010
Leverandør: Thermo Fisher Scientific
CAS nummer: 1120-71-4
Gå til:
Product Details & Documents
Synonymer: 1,2-Oxathiolane 2,2-dioxide
- CAS nummer:1120-71-4
- Formel:C₃H₆O₃S
- MDL nummer:MFCD00005355
- Molecular Weight:122.14 g/mol
- Kogende Pt:180 °C (30 mmHg)
- Smeltende Pt:30…33 °C
- Tæthed:1,392 g/cm³
- Flash Pt:>110 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:214-317-9
- FN:2811
- ADR:6.1,III
Documents
ACRO13142
SDS
Specifikationer
Specifikationstestresultater
- Appearance:White to light yellow Low melting solid or liquid
- Infrared spectrum:Conforms
- Melting point:28 to 33 °C
- GC:≥96.0 %
Specifikationer
- Pakning:Glass bottle
- Pakn:1 kg



