Orders LinkContactUsLinkComponent
Electronic overhead stirrers, MICROSTAR control
Electronic overhead stirrers, MICROSTAR control
MICROSTAR 15 control overhead stirrer
  441-0465
 :   IKA
Electronic overhead stirrers, MICROSTAR control
  441-0465
 :  IKA
Electronic overhead stirrers, MICROSTAR control
Electronic overhead stirrers, MICROSTAR control
MICROSTAR 15 control overhead stirrer
12.020,00 kr
1 * 1 Items
These overhead stirrers are ideal for special applications. Combining high performance with excellence, the units require minimum space.
  • Fast, responsive display for maximum visibility and chemical resistance
  • Clear display shows all essential information at a glance
  • Integrated timer/counter for the control of kinetic sensitive reactions and reminders
  • USB interface for documenting parameters using labworldsoft® or installing firmware updates
  • Intuitive and simple handling; touch-sensitive surface for long service life
  • Temperature reading is shown on display
  • Chemical resistant housing is enclosed in hardened glass
  • Key lock function
Microprocessor-controlled speed monitoring provides constant rotational speed, even with changes in viscosity. The state-of-art vibration sensors detect deviations from permissible thresholds and automatically stop the process.
Temperature measurement range: −10 to +350 °C.
Certifications: IP protection class according to DIN EN 60529: IP 54
  • Beskrivelse:
    MICROSTAR 15 control overhead stirrer
  • Seq:
    02
  • Maks. omrøringskapacitet:
    10 L
  • Maks. viskositet:
    8000 mPas
  • Plug-in tilkobling Ø:
    32/22 mm
  • Hastighedsområde I:
    30 - 1000 min⁻¹
  • Vandringslængde:
    0,5 - 8,0 mm
  • Permissible relative humidity:
    80%
  • Cat. No.:
    441-0465
  • Effekt - Motorindgang/-udgang:
    32/22 W
  • Vægt:
    1,26 kg
  • Model:
    MICROSTAR 15 control
  • W×D×H:
    60×110×123 mm
  • Maks. moment ved patron:
    15 Ncm
  • Permissible ambient temperature:
    5…40 °C
Accessories
  •  
    Beskrivelse
  • IKAA20118833
    H 71.45 Temperature sensor 450 mm, PTFE coated, Ø 8 mm
    1 * 1 Items - 5.360,00 kr
  •  
    Beskrivelse
  • 620-8104
    Forlængerkabel, til adskillelse mellem hus og sensor for at undgå skade fra dampe afgivet af mediet
    1 * 1 Items - 1.980,00 kr
  •  
    Beskrivelse
    Omrører - Ø
    Skaft - Ø
  • 441-2951
    Dispergeringsomrører
    80 mm
    8 mm
    1 * 1 Items - 2.650,00 kr
  •  
    Beskrivelse
  • IKAA25004318
    IKA [scale] Weighing stand, capacity 5 to 6000 g
    1 * 1 Items - 8.000,00 kr
  •  
    Beskrivelse
    Omrører - Ø
    Skaft - Ø
  • 441-2962
    Centrifugal omrører
    100/24 mm
    8 mm
    1 * 1 Items - 2.230,00 kr
  •  
    Beskrivelse
    Til
  • IKAA25007821
    R 60.1 Keyless chuck
    use with MINISTAR 20/40/80 digital and control overhead stirrers
    1 * 1 Items - 795,00 kr
  •  
    Beskrivelse
  • 431-2944
    Båndklemme til sikring af beholdere, stativ Ø 25 - 36 mm, beholder Ø 40 - 300 mm
    1 * 1 Items - 3.570,00 kr
  •  
    Beskrivelse
    Omrører - Ø
    Skaft - Ø
  • 441-2954
    Dispergeringsomrører
    42 mm
    8 mm
    1 * 1 Items - 4.120,00 kr
  •  
    Beskrivelse
    Til
  • IKAA20019397
    Labworldsoft® 6 Starter
    Small configurations with up to 3 devices can be controlled at the same time
    1 * 1 Items - 45.630,00 kr
  •  
    Beskrivelse
    Omrører - Ø
    Skaft - Ø
  • 441-2912
    Propelomrører, tre blade
    55 mm
    8 mm
    1 * 1 Items - 3.110,00 kr
  •  
    Beskrivelse
  • 442-0316
    Glasbelagt sensor til brug med ETS-D5 og VT-5 kontakttermometre
    1 * 1 Items - 2.850,00 kr
  •  
    Beskrivelse
    Til
    Model
  • 441-2129
    Pladestativ, 200×316 mm, stang 16 mm Ø, højde 1000 mm
    T 18 basic, T 25 digital
    Pladestativ R 1827
    1 * 1 Items - 3.340,00 kr
  •  
    Beskrivelse
    Omrører - Ø
    Skaft - Ø
  • 441-2901
    Omrører, fire blade
    50 mm
    8 mm
    1 * 1 Items - 1.490,00 kr
  •  
    Beskrivelse
    Til
    Model
  • 431-2901
    Pladestativ, 200×316 mm, stang 16 mm Ø, højde 800 mm
    T 18 basic, T 25 digital
    Pladestativ R 1826, plade 200x316 mm, stang 800 mm
    1 * 1 Items - 3.130,00 kr
  •  
    Beskrivelse
  • 431-1047
    Plate stand R 1825, 200×316 mm, rod 16 mm Ø with height 560 mm
    1 * 1 Items - 2.840,00 kr
  •  
    Beskrivelse
  • 441-9213
    Fleksibel kobling til applikationer, der anvender glasomrøringsstave, Ø 6 - 10 mm, maks. drejningsmoment 10 Ncm
    1 * 1 Items - 3.010,00 kr
  •  
    Beskrivelse
    Omrører - Ø
    Skaft - Ø
  • IKAA20001203
    Spiral stirrer
    50 mm
    8 mm
    1 * 1 Items - 3.340,00 kr
  •  
    Beskrivelse
    Til
  • 431-0004
    T 25, stativ med Ø på 25 - 36 mm, forlængerarm med Ø på 5 - 21 mm
    T 50 basic
    1 * 1 Items - 1.180,00 kr
  •  
    Beskrivelse
    Omrører - Ø
    Skaft - Ø
  • 441-2911
    Propelomrører, tre blade
    45 mm
    8 mm
    1 * 1 Items - 2.980,00 kr
  •  
    Beskrivelse
  • 431-2912
    Pneumatisk teleskopstativ, 460×420 mm, stang 34 mm Ø, variabel højde 620 - 1010 mm
    1 * 1 Items - 12.600,00 kr
  •  
    Beskrivelse
  • IKAA25006431
    R 1346 (PTFE-coated) Propeller stirrer, 4-bladed, 350 mm shaft length, Stirrer Ø 58 mm, Shaft Ø: 8 mm (uncoated part Ø: 6 mm)
    1 * 1 Items - 2.440,00 kr
  •  
    Beskrivelse
  • 441-2999
    Beskyttelsesholder
    1 * 1 Items - 1.790,00 kr
  •  
    Beskrivelse
    Omrører - Ø
    Skaft - Ø
  • 441-2961
    Centrifugal omrører
    60/15 mm
    8 mm
    1 * 1 Items - 1.680,00 kr
  •  
    Beskrivelse
    Omrører - Ø
    Skaft - Ø
  • 441-2931
    Ankeromrører
    45 mm
    8 mm
    1 * 1 Items - 3.790,00 kr
  •  
    Beskrivelse
    Omrører - Ø
    Skaft - Ø
  • 441-2903
    Omrører, fire blade
    100 mm
    8 mm
    1 * 1 Items - 1.830,00 kr
  •  
    Beskrivelse
  • 441-2970
    Stainless steel screw type stirrer
    1 * 1 Items - 1.350,00 kr
  •  
    Beskrivelse
  • 442-0252
    Ekstra sensor af rustfrit stål til brug sammen med ETS-D5
    1 * 1 Items - 1.960,00 kr
  •  
    Beskrivelse
  • 431-9114
    H-stativ, 460×420 mm, stang 34 mm Ø med højde på 1010 mm
    1 * 1 Items - 6.800,00 kr
  •  
    Beskrivelse
    Omrører - Ø
    Skaft - Ø
  • 441-2932
    Ankeromrører
    90 mm
    8 mm
    1 * 1 Items - 4.580,00 kr
  •  
    Beskrivelse
    Til
    Model
  • 431-2943
    Båndklemme til sikring af beholdere, stativ Ø 8 - 16 mm, beholder Ø 40 - 300 mm
    T 18 basic, T 25 digital
    Klemmeenhed RH 3
    1 * 1 Items - 2.900,00 kr
  •  
    Beskrivelse
    Til
  • IKAA20017366
    Labworldsoft® 6 Pro, with access to the entire device library
    All configurations with any number of devices can be controlled at the same time
    1 * 1 Items - 104.110,00 kr
  •  
    Beskrivelse
    Omrører - Ø
    Skaft - Ø
  • 441-2942
    Padleformet omrører
    70 mm
    8 mm
    1 * 1 Items - 1.790,00 kr
  •  
    Beskrivelse
  • IKAA20003417
    Beaker cap, BC 1000, silicone, for 600 and 1000 ml beakers
    1 * 1 Items - 180,00 kr
  •  
    Beskrivelse
  • IKAA20117772
    H 71.32 Temperature sensor 320 mm, PTFE coated, Ø 8 mm
    1 * 1 Items - 4.650,00 kr
  •  
    Beskrivelse
  • 441-0038
    Støtteholder til omrøreraksel
    1 * 1 Items - 2.130,00 kr
  •  
    Beskrivelse
    Omrører - Ø
    Skaft - Ø
  • 441-2125
    Omrører, tre blade (PTFE)
    75 mm
    8 mm
    1 * 1 Items - 5.380,00 kr
  •  
    Beskrivelse
  • 441-2120
    Spare stainless steel paddle stirrer
    1 * 1 Items - 1.580,00 kr
  •  
    Beskrivelse
  • 441-0344
    Temperature sensor, coated
    1 * 1 Items - 3.430,00 kr
  •  
    Beskrivelse
    Omrører - Ø
    Skaft - Ø
  • 441-2922
    Turbineomrører
    50 mm
    8 mm
    1 * 1 Items - 2.570,00 kr
  •  
    Beskrivelse
    Omrører - Ø
    Skaft - Ø
  • 441-2921
    Turbineomrører
    30 mm
    8 mm
    1 * 1 Items - 2.010,00 kr
  •  
    Beskrivelse
  • IKAA20001206
    Radial stirrer, R 3004, shaft (Ø×L) 8,0×359 mm, stirrer Ø 30 mm
    1 * 1 Items - 2.900,00 kr
  •  
    Beskrivelse
    Til
    Model
  • 431-2923
    Dobbeltmuffe, 6 - 16 mm Ø stativ, 6 - 16 mm Ø forlængerarm
    T 18 basic, T 25 digital
    Klemme R 182
    1 * 1 Items - 902,00 kr