Orders LinkContactUsLinkComponent
Graphite tubes for atomic absorption
Graphite tubes for atomic absorption
Pyrolytically coated partition graphite tube for extended life
Katalog # PERCN9304627
Leverandør:   PerkinElmer
Graphite tubes for atomic absorption
Katalog # PERCN9304627
Leverandør:  PerkinElmer
Graphite tubes for atomic absorption
Pyrolytically coated partition graphite tube for extended life
Price
9.360,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Graphite tubes made with highest quality material.
Suitable AA instruments.
Specifikationer
  • Beskrivelse:
    Pyrolytically coated partition graphite tube for extended life
  • Seq:
    505