Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Accessories for Sampler, Vampire (Bürkle)

Accessories for Sampler, Vampire (Bürkle)

Basket for 500 ml sample bottles, 84 mm
Catalog # BURK5302-2051
Supplier:   BURKLE
Accessories for Sampler, Vampire (Bürkle)
Catalog # BURK5302-2051
Supplier:  BURKLE
Accessories for Sampler, Vampire (Bürkle)

Basket for 500 ml sample bottles, 84 mm

Price
757,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Beskrivelse:
    Basket for 500 ml sample bottles, 84 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.