n-Hexan, anhydrous (max. 0.003% H₂O)
Glass bottle with septum cap, 250 mL
Leverandør: VWR Chemicals
n-Hexan, anhydrous (max. 0.003% H₂O)
Katalog # 85819.230
Leverandør: VWR Chemicals
CAS nummer: 110-54-3
Gå til:
Product Details & Documents
Synonymer: Hexyl hydride, Hexanen, Hexane
- CAS nummer:110-54-3
- Formel:H₃C(CH₂)₄CH₃
- MDL nummer:MFCD00009520
- Molecular Weight:86.18 g/mol
- Merck Index:13,04712
- Kogende Pt:69 °C (1013 hPa)
- Smeltende Pt:–94,3 °C
- Tæthed:0,659 g/cm³ (20 °C)
- Flash Pt:–22 °C
- Opbevaringstemperatur:Stue temperatur
- FN:1208
- ADR:3,II
- :0,2 µm Filtreret. Packaged under inert gas.
- EINECS:203-777-6
Documents
Specifikationer
Specifikationstestresultater
- Assay (GC, on anhydrous basis):99-100 %
- Color (APHA, Pt-Co):0-10
- Water (KF):0-0.003 % w/w
- Acidity (as Acetic acid):0-0.001 %
- Residue after evaporation:0-0.0005 % w/w
Specifikationer
- Pakning:Glass bottle with septum cap
- Pakn:250 mL
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



