Orders LinkContactUsLinkComponent
Accessories for pipette controller, MACROMAN™
Mains adapter (100 - 240 VAC, 50/60 Hz) with 4 plugs (EU, UK, AUS, US)
Katalog # GILSF1077527
Leverandør:   GILSON
Accessories for pipette controller, MACROMAN™
Katalog # GILSF1077527
Leverandør:  GILSON
Accessories for pipette controller, MACROMAN™
Accessories for pipette controller, MACROMAN™
Mains adapter (100 - 240 VAC, 50/60 Hz) with 4 plugs (EU, UK, AUS, US)
Price
883,26 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Mains adapter (100 - 240 VAC, 50/60 Hz) with 4 plugs (EU, UK, AUS, US)
  • Seq:
    07