Orders LinkContactUsLinkComponent
End point detection system, End-Zone™
End point detection system, End-Zone™
End-Zone™ starter kit
Katalog # 535-0392
Leverandør:   LABCONCO
End point detection system, End-Zone™
Katalog # 535-0392
Leverandør:  LABCONCO
End point detection system, End-Zone™
End point detection system, End-Zone™
End-Zone™ starter kit
Price
12.670,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
End-Zone™ provides an alternative to visual inspection making it easy to determine when freeze drying is complete.
  • Alerts when sample reaches the user-selected moisture level
  • Works with FreeZone Lyo-Works™ OS
Starter kit is used for detecting end point in one sample flask. Add-on Kit is used for detecting end point in up to five sample.
End-Zone™ will alert when sample moisture levels have reached the user-selected level. It compares the vacuum level of an empty valve to the vacuum level inside a fast-freeze flask containing the sample.
Leveringsinformation: Starter Kit includes two vacuum sensors, one valve adapter, one fast-freeze flask top with sensor port, a connection cable, and a mountable holder with space for six sensors. Add-on kit includes one vacuum sensor and one Fast-Freeze flask top with an opening to insert the vacuum sensor.
Specifikationer
  • Beskrivelse:
    End-Zone™ starter kit
  • Seq:
    10