Cleaning coupons
Cleaning coupon, 304 stainless steel (bright polish), 100×100 mm
Katalog # SAMP8644S4-100B
Leverandør: Sampling Systems
Gå til:
Product Details & Documents
These cleaning validation coupons are available in all popular materials and various sizes.
Specifikationer
- Beskrivelse:Cleaning coupon, 304 stainless steel (bright polish)
- L×B:100×100 mm
- Seq:03