Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
WHEATON® FTA® Nucleic Acid Collection Storage Boxes

WHEATON® FTA® Nucleic Acid Collection Storage Boxes

457,2×177,8×101,6 mm
Catalog # 479-2127
Supplier:   DWK Life Sciences
WHEATON® FTA® Nucleic Acid Collection Storage Boxes
Catalog # 479-2127
Supplier:  DWK Life Sciences
WHEATON® FTA® Nucleic Acid Collection Storage Boxes

457,2×177,8×101,6 mm

Price
758,00 kr
Log in for availability
1 * 4 Items
Product Details & Documents

Storage boxes provide an ideal method for batching/storing FTA® Nucleic acid collection pouches.

  • Water repellent fiberboard box material allows for longer durability
  • For use with small or large FTA cards
  • Accommodates two rows of 76,2 mm FTA card pouches or one row of 152,4 FTA card pouches
  • Removable divider for use with smaller pouches
  • For use at RT, +4, −20 °C or colder

Storage boxes are ideal for nucleic acid collection.

Specifications
  • Seq:
    02
  • B×D×H:
    457,2×177,8×101,6 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.