Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

WHEATON® FTA® Nucleic Acid Collection Storage Boxes

457,2×152,4×101,6 mm
Katalog # 479-2126
Leverandør:   DWK Life Sciences
WHEATON® FTA® Nucleic Acid Collection Storage Boxes
Katalog # 479-2126
Leverandør:  DWK Life Sciences
WHEATON® FTA® Nucleic Acid Collection Storage Boxes

457,2×152,4×101,6 mm

Price
390,00 kr
Log ind for tilgængelighed
1 * 2 Items
Product Details & Documents

Storage boxes provide an ideal method for batching/storing FTA® Nucleic acid collection pouches.

  • Water repellent fiberboard box material allows for longer durability
  • For use with small or large FTA cards
  • Accommodates two rows of 76,2 mm FTA card pouches or one row of 152,4 FTA card pouches
  • Removable divider for use with smaller pouches
  • For use at RT, +4, −20 °C or colder

Storage boxes are ideal for nucleic acid collection.

Specifikationer
  • Seq:
    01
  • B×D×H:
    457,2×152,4×101,6 mm