Orders LinkContactUsLinkComponent
Gas filter connecting units
Gas filter connecting units
Gas filter connecting unit with 4-position, fitting size 1/4"
Katalog # SGEA1035044
Gas filter connecting units
Katalog # SGEA1035044
Gas filter connecting units
Gas filter connecting unit with 4-position, fitting size 1/4"
Price
8.570,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
A connecting unit with inlet and outlet connectors for the gas lines.
  • Allow easy installation of gas purifiers into the gas line
  • Unit can be bench or wall mounted
  • Available in 1,2 and 4-port configurations
Gas purifiers are an essential part of your GC analysis as contaminants in gases can significantly impact the quality of results. Oxygen, hydrocarbons and moisture can lead to problems such as noisy baselines, moisture entering the GC column, excessive bleed and septa degradation. Even if carrier gas is of the highest quality, contaminants can be picked up from every part of the gas line. Therefore, a gas purifier is needed to ensure that maximum productivity is achieved.
Ordering information: Connecting units and filters must be ordered separately.
Documents
Specifikationer
  • Beskrivelse:
    Gas filter connecting unit with 4-position, fitting size 1/4"
  • Seq:
    07