Coats, paint-tex® plus
hvid, XXL
Katalog # ZVGA30775-00
Leverandør: Zellstoff-Vertriebs-GmbH & Co. KG
Gå til:
Product Details & Documents
Suitable for longer use due to the breathable SMS material.
- Features snap buttons
- Collar and cuffs
- Three pockets
Ideal for renovation work, in the remediation of contaminated sites and asbestos.
Certifications: Cat. III/CE 1023/Type 5 + 6
Specifikationer
- Tøj størrelse:XXL
- Farve:hvid
- Seq:03
- Cat. No.:ZVGA30775-00
- Outfit applikationer:Industrial
- Lukkestørrelse:Med trykknapper
- Lommer:med lommer