Rain hat, 9920 RS
fluorescerende gul, M/L
Katalog # KWIN114043130M/L
Leverandør: FRISTADS KANSAS
Gå til:
Product Details & Documents
This rain hat is made from PU-coated polyester jersey and features a fleece lining (polyester).
- Ear flaps with drawstring
- Bright colour for improved visibility
Weight: 170 g/m²
Certifications: EN 343 (3-1)
Documents
Specifikationer
- Tøj størrelse:M/L
- Farve:fluorescerende gul
- Seq:01
- Køn:Unisex
- Cat. No.:KWIN114043130M/L
- Risk area:Rain (waterproof)
- Vægt:170 g/m²
- Model:9920 RS
- Outfit applikationer:Outdoor
- Stof:PU-coated polyester