Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Mouse Recombinant IL-4

Mouse Recombinant IL-4

20 µG
Katalog # STMC78047.1
Marketsource
Leverandør:   Stemcell Technologies
Mouse Recombinant IL-4
Katalog # STMC78047.1
Leverandør:  Stemcell Technologies
Marketsource
CAS nummer:  
Mouse Recombinant IL-4
Mouse Recombinant IL-4

20 µG

CAS nummer:  
Price
2.078,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Interleukin 4 (IL-4) is a type I cytokine produced by Th2 T cells, NK T cells, γ/δ T cells, eosinophils, mast cells, and activated basophils. IL-4 receptors are present on a number of cells, including hematopoietic, endothelial, epithelial, muscle, and brain tissues. IL-4 receptor engagement initiates signaling through the JAK/STAT pathway, and leads to the activation of PI3K and Ras/MAPK pathways (Nelms et al.). IL-4 plays a key role in immunoglobulin class switching in B cells and in regulating the differentiation of naïve T cells into T helper 2 (Th2) cells, while inhibiting Th1 differentiation. IL-4 acts together with tumor necrosis factor to stimulate expression of adhesion molecules on vascular endothelial cells and to downregulate expression of E-selectin, thus recruiting T cells and eosinophils to the site of inflammation (Nelms et al.).

Advarsel: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifikationer

Specifikationer

  • Pakn:
    20 µG
  • Cat. No.:
    STMC78047.1
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    IL-4
  • Renhed:
    ≥95%
  • Species:
    Mouse

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.