Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Mouse Recombinant IL-6

Mouse Recombinant IL-6

1.000 µG
Katalog # STMC78052.2
Marketsource
Leverandør:   Stemcell Technologies
Mouse Recombinant IL-6
Katalog # STMC78052.2
Leverandør:  Stemcell Technologies
Marketsource
CAS nummer:  
Mouse Recombinant IL-6
Mouse Recombinant IL-6

1.000 µG

CAS nummer:  
Price
33.654,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Interleukin 6 (IL-6) is a pleiotropic growth factor with a wide range of biological activities in immune regulation, hematopoiesis, and oncogenesis. IL-6 is produced by a variety of cell types including T cells, B cells, monocytes and macrophages, fibroblasts, hepatocytes, vascular endothelial cells, and various tumor cell lines. On its own or in combination with other factors such as IL-2 and interferon-γ, IL-6 stimulates the proliferation of B cells, T cells, and hybridoma cells (Nordan et al.; Van Snick et al.; Gauldie et al.; Mihara et al.; Tanaka et al). In combination with cytokines such as IL-3, GM-CSF, and SCF, IL-6 has been shown to promote hematopoietic progenitor cell proliferation and differentiation in vitro. IL-6 signals through a cell surface type I cytokine receptor complex consisting of the ligand-binding IL-6α (CD126) and the signal-transducing gp130 subunits. The binding of IL-6 to its receptor system includes activation of the JAK/STAT signaling pathway (Mihara et al.; Peters et al; Tanaka et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifikationer

Specifikationer

  • Pakn:
    1.000 µG
  • Cat. No.:
    STMC78052.2
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    IL-6
  • Renhed:
    ≥95%
  • Species:
    Mouse

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.