Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Mouse Recombinant LIF

Mouse Recombinant LIF

1.000 µG
Katalog # STMC78056.2
Marketsource
Leverandør:   Stemcell Technologies
Mouse Recombinant LIF
Katalog # STMC78056.2
Leverandør:  Stemcell Technologies
Marketsource
CAS nummer:  
Mouse Recombinant LIF
Mouse Recombinant LIF

1.000 µG

CAS nummer:  
Price
31.421,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Leukemia inhibitory factor (LIF) is an interleukin 6 class cytokine that regulates a broad variety of developmental functions. After LIF binds to the LIF receptor (LIFR), LIFR associates with gp130 and activates JAK/STAT and MAPK signaling (Auernhammer and Melmed; Suman et al.). LIFR activation of STAT3 is essential for maintaining the mouse embryonic stem cell phenotype (Niwa et al.). Produced by the endometrium, LIF plays an important autocrine and paracrine role in implantation by regulating proliferation, invasion, and differentiation of trophoblasts following blastocyst attachment (Auernhammer and Melmed; Suman et al.). Human LIF can be used for the maintenance of mouse embryonic stem cells; however, mouse LIF cannot bind to the human receptor, thus rendering mouse LIF inactive (Dahéron et al.). LIF is produced by CD4+ and activated regulatory T cells, and promotes Foxp3 expression, while repressing Th17 lineage-specific genes (Metcalfe). LIF is also secreted by mesenchymal stromal cells, where it supports hematopoiesis and immune modulation (Nasef et al.).

 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifikationer

Specifikationer

  • Pakn:
    1.000 µG
  • Cat. No.:
    STMC78056.2
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    LIF
  • Renhed:
    ≥95%
  • Species:
    Mouse

Specifikationer