Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Mouse Recombinant SCF (<i>E. coli</i>-expressed)

Mouse Recombinant SCF (E. coli-expressed)

10 µG
Katalog # STMC78064.1
Marketsource
Leverandør:   Stemcell Technologies
Mouse Recombinant SCF (<i>E. coli</i>-expressed)
Katalog # STMC78064.1
Leverandør:  Stemcell Technologies
Marketsource
CAS nummer:  
Mouse Recombinant SCF (<i>E. coli</i>-expressed)
Mouse Recombinant SCF (E. coli-expressed)

10 µG

CAS nummer:  
Price
1.731,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Stem cell factor (SCF) is an early-acting cytokine that plays a pivotal role in the regulation of embryonic and adult hematopoiesis. SCF promotes cell survival, proliferation, differentiation, adhesion, and functional activation of cells at multiple levels of the hematopoietic hierarchy. Together with other cytokines such as thrombopoietin and Flt3/Flk-2 Ligand, SCF is commonly used to promote expansion of primitive hematopoietic stem cells and multi-potent progenitor cells in culture (Huang et al.; Kent et al.). In synergy with various growth factors, including IL-2, IL-3, IL-6, IL-7, G-CSF, and erythropoietin, SCF increases proliferation and differentiation of myeloid and erythroid progenitor cells and a subset of lymphoid progenitor cells (Broudy). In the mouse, SCF is essential during fetal gonadal development (Mauduit). It is produced by stromal cells in the fetal liver, bone marrow, and thymus, in the central nervous system, in keratinocytes, and in the gut mucosa, and can function as a chemotactic and chemokinetic factor. SCF exists in two biologically active splice forms: a soluble and a transmembrane isoform. Upon binding to its receptor (c-kit tyrosine kinase receptor; CD117), it activates PI3K, JAK/STAT, and MAPK pathways. SCF and signaling from c-kit has also been reported to play an important role in pigmentation, fertility, vasculogenesis, motility of the gut via c-kit-positive interstitial cells of Cajal, and in the migration of neuronal stem and progenitor cells to sites of injury in the brain (Lennartsson and Ronnstrand).

Advarsel: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifikationer

Specifikationer

  • Pakn:
    10 µG
  • Cat. No.:
    STMC78064.1
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    SCF
  • Protein synonyms:
    Kit ligand|Mast cell growth factor|Steel factor|Stem cell factor
  • Renhed:
    ≥95%
  • Kilde:
    E. coli
  • Species:
    Mouse

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.