Order Entry
Denmark
Orders LinkContactUsLinkComponent
Mouse Recombinant IL-11

Mouse Recombinant IL-11

10 µG
Katalog # STMC78026.1
Marketsource
Leverandør:   Stemcell Technologies
Mouse Recombinant IL-11
Katalog # STMC78026.1
Leverandør:  Stemcell Technologies
Marketsource
CAS nummer:  
Mouse Recombinant IL-11
Mouse Recombinant IL-11

10 µG

CAS nummer:  
Price
1.844,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Interleukin 11 (IL-11) is a pleiotropic cytokine with effects on various tissues including the bone marrow, brain, and intestinal mucosa (Du and amp; Williams). It belongs to the IL-6 family of cytokines that share a common signal transducer, gp130. Culture of mouse bone marrow cells with IL-11 in combination with IL-3, IL-6, and stem cell factor induces significant expansion and proliferation of colony-forming cells in vitro (Peters et al.). In addition, in combination with IL-3, IL-11 significantly enhances the growth of megakaryocytic colonies in vitro, suggesting its role in augmenting mouse megakaryopoiesis (Yonemura et al.). IL-11 is expressed in a wide range of normal adult mouse tissues, including the central nervous system, thymus, lung, and bone. The mouse IL-11 cDNA was cloned using an expression library generated from the lipopolysaccharide-induced mouse fetal thymic cell line, T2 (Morris et al.). The binding of IL-11 to its receptor induces heterodimerization with the gp130 subunit and activation of JAK tyrosine kinases. IL-11 also plays a role in cancer progression by inducing the proliferation of epithelial cancer cells and the survival of metastatic cells at distant organs. Recently, IL-11 has gained interest for its role in the pathogenesis of diseases in dysregulated mucosal homeostasis associated with STAT3 upregulation, including gastrointestinal cancers (Putoczki et al.).

Advarsel: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifikationer

Specifikationer

  • Pakn:
    10 µG
  • Cat. No.:
    STMC78026.1
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    IL-11
  • Renhed:
    ≥95%
  • Species:
    Mouse

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.