Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Human Recombinant IL-17A

25 µG
Katalog # STMC78032.1
Marketsource
Leverandør:   Stemcell Technologies
Human Recombinant IL-17A
Katalog # STMC78032.1
Leverandør:  Stemcell Technologies
Marketsource
CAS nummer:  
Human Recombinant IL-17A
Human Recombinant IL-17A

25 µG

CAS nummer:  
Price
2.078,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Interleukin 17A (IL-17A) is the founding member of the family of cytokines that includes IL-17B through IL-17F. It is a potent pro-inflammatory cytokine that plays a key role in defense against pathogens. IL-17A and IL-17F signal as homodimers or heterodimers through the same receptor, and activate NF-κB, MAPK, and C/EBP pathways (Gaffen). IL-17A is produced by Th17 cells, CD8+ T cells, γ/δ T cells, natural killer (NK) T cells, B cells, innate lymphoid cells, and mesenchymal stromal cells (MSCs) (Cua and Tato; Gaffen; Mojsilović et al.). IL-17A mediates protection against extracellular pathogens, and together with IL-22 stimulates production of antimicrobial peptides. It induces granulopoiesis factors and neutrophil-specific chemokines. Together with tumor necrosis factor alpha (TNF-α), IL-17A induces a sustained neutrophil recruitment during inflammation (Cua and Tato). IL-17A receptor is expressed at particularly high levels on stromal cells, including MSCs. IL-17A increases the frequency and the average size of fibroblast colony-forming units (CFU-F), as well as the proliferation of marrow-derived MSCs. It enhances osteogenic differentiation, and inhibits adipocyte differentiation and chondrogenesis (Mojsilović et al.).

 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifikationer

Specifikationer

  • Pakn:
    25 µG
  • Cat. No.:
    STMC78032.1
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    IL-17A
  • Renhed:
    ≥95%
  • Species:
    Human

Specifikationer