Orders LinkContactUsLinkComponent
EasySep™ Mouse CD49b Positive Selection Kits
EasySep™ mouse CD49b positive selection kit, 2×10⁹ cells
Katalog # STMC18755
Marketsource
Leverandør:   Stemcell Technologies
EasySep™ Mouse CD49b Positive Selection Kits
Katalog # STMC18755
Leverandør:  Stemcell Technologies
Marketsource
EasySep™ Mouse CD49b Positive Selection Kits
EasySep™ Mouse CD49b Positive Selection Kits
EasySep™ mouse CD49b positive selection kit, 2×10⁹ cells
Price
5.989,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Immunomagnetic positive selection of CD49b+ cells (e.g. NK cells) from mouse splenocytes or other tissues.
  • Fast and easy-to-use
  • Up to 90% purity
  • No columns required
The EasySep™ Mouse CD49b Positive Selection Kit is designed to isolate CD49b+ cells from single-cell suspensions of splenocytes or other tissues by positive selection. The cocktail contains a combination of PE-labelled antibodies that is directed against CD49b, which are then recognized by Tetrameric Antibody Complexes that are directed against PE and dextran. labelled cells are bound to magnetic particles and separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.
 : Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifikationer
  • Beskrivelse:
    EasySep™ mouse CD49b positive selection kit, 2×10⁹ cells
  • Seq:
    01