Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Collagenase B, ACF

Collagenase B, ACF

Collagenase B, ACF, 1 g
Katalog # STMC07439
Marketsource
Leverandør:   Stemcell Technologies
Collagenase B, ACF
Katalog # STMC07439
Leverandør:  Stemcell Technologies
Marketsource
CAS nummer:  
Collagenase B, ACF
Collagenase B, ACF

Collagenase B, ACF, 1 g

CAS nummer:  
Price
5.651,00 kr
Log ind for tilgængelighed
1 * 1 g
Product Details & Documents

Animal component-free collagenase for the digestion of native collagen fibrils.

  • Obtain cleaner cultures with an animal component-free solution

Use Collagenase B, Animal Component-Free (ACF) to digest native collagen fibrils in connective tissues and prevent the introduction of animal-derived pathogens. This enzymatic preparation is sourced from Clostridium histolyticum and is obtained from cultures free of animal-derived materials. Collagenase preparations contain the activity of several proteases, including collagenase, caseinase, clostripain, and trypsin (Kessler and Yaron). Collagenase B, ACF contains higher collagenase and caseinase levels than Collagenase A.

Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifikationer

Specifikationer

  • Pakn:
    1 g
  • Beskrivelse:
    Collagenase B, ACF
  • Cat. No.:
    STMC07439
  • Species:
    Non-human primate|Human|Rat|Mouse

Specifikationer