Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Mouse Recombinant IL-1 beta

Mouse Recombinant IL-1 beta

50 µG
Katalog # STMC78035
Marketsource
Leverandør:   Stemcell Technologies
Mouse Recombinant IL-1 beta
Katalog # STMC78035
Leverandør:  Stemcell Technologies
Marketsource
CAS nummer:  
Mouse Recombinant IL-1 beta
Mouse Recombinant IL-1 beta

50 µG

CAS nummer:  
Price
5.339,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Interleukin 1 beta (IL-1β) is synthesised as an inactive precursor protein or pro-IL-1β. This precursor is cleaved intracellularly by caspase 1 (IL-1β convertase) to form the active form of the protein that is later secreted (Allan et al.). IL-1β binds to IL-1 receptor and activates intracellular signaling via the MAPK or NF-kB pathway. IL-1β is released by monocytes, tissue macrophages, and dendritic cells in response to infection or injury and induces expression of acute-phase proteins. It also promotes the infiltration of inflammatory and immunocompetent cells from the circulation into the extravascular space and affected tissues, by stimulating the expression of adhesion molecules on endothelial cells. IL-1β also affects other immune cells; for example, it co-stimulates T cell functions together with antigen or mitogen. It also stimulates Th17 differentiation and B cell proliferation in an IL-6-dependent manner. Mice deficient in IL-1β do not show phenotypical differences from wild-type mice; however, they have a reduced response to inflammation, suggesting that IL-1β plays a key role in inflammatory diseases (Dinarello).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifikationer

Specifikationer

  • Pakn:
    50 µG
  • Cat. No.:
    STMC78035
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    IL-1 beta
  • Renhed:
    ≥95%
  • Species:
    Mouse

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.