Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Mouse Recombinant Shh

Mouse Recombinant Shh

50 µG
Katalog # STMC78066
Marketsource
Leverandør:   Stemcell Technologies
Mouse Recombinant Shh
Katalog # STMC78066
Leverandør:  Stemcell Technologies
Marketsource
CAS nummer:  
Mouse Recombinant Shh
Mouse Recombinant Shh

50 µG

CAS nummer:  
Price
3.269,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Sonic hedgehog (Shh) is a member of the Hedgehog family of secreted signaling proteins that has a prominent role in patterning during early mammalian development (Ho and Scott). It is expressed during early embryogenesis in a variety of tissues (Finco et al.). During patterning of the central nervous system, Shh directly acts on cells to specify neural cell fates, regulates proliferation and survival of oligodendrocyte precursors and neural crest cells and stimulates proliferation, differentiation, and axon growth (Ho and Scott; Martí). Shh also regulates T cell differentiation and activation (Crompton et al.). Recombinant Mouse Shh is a fully biologically active molecule.

Advarsel: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifikationer

Specifikationer

  • Pakn:
    50 µG
  • Cat. No.:
    STMC78066
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    Shh
  • Renhed:
    ≥95%
  • Species:
    Mouse

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.