Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Collagenase B, ACF

Collagenase B, ACF

Collagenase B, ACF, 100 mg
Katalog # STMC07438
Marketsource
Leverandør:   Stemcell Technologies
Collagenase B, ACF
Katalog # STMC07438
Leverandør:  Stemcell Technologies
Marketsource
CAS nummer:  
Collagenase B, ACF
Collagenase B, ACF

Collagenase B, ACF, 100 mg

CAS nummer:  
Price
1.233,00 kr
Log ind for tilgængelighed
1 * 100 mg
Product Details & Documents

Animal component-free collagenase for the digestion of native collagen fibrils.

  • Obtain cleaner cultures with an animal component-free solution

Use Collagenase B, Animal Component-Free (ACF) to digest native collagen fibrils in connective tissues and prevent the introduction of animal-derived pathogens. This enzymatic preparation is sourced from Clostridium histolyticum and is obtained from cultures free of animal-derived materials. Collagenase preparations contain the activity of several proteases, including collagenase, caseinase, clostripain, and trypsin (Kessler and Yaron). Collagenase B, ACF contains higher collagenase and caseinase levels than Collagenase A.

Advarsel: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifikationer

Specifikationer

  • Pakn:
    100 mg
  • Beskrivelse:
    Collagenase B, ACF
  • Cat. No.:
    STMC07438
  • Species:
    Non-human primate|Human|Rat|Mouse

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.