Order Entry
Denmark
Orders LinkContactUsLinkComponent
Mouse Recombinant IL-2

Mouse Recombinant IL-2

20 µG
Catalog # STMC78081
Marketsource
Mouse Recombinant IL-2
Catalog # STMC78081
Marketsource
CAS Number:  
Mouse Recombinant IL-2
Mouse Recombinant IL-2

20 µG

CAS Number:  
Price
1.710,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents

Interleukin 2 (IL-2) is a monomeric cytokine that was originally identified as a T cell growth factor (Gaffen and Liu). It binds to a heterotrimeric receptor consisting of CD25, CD122, and CD132. Upon binding, it activates JAK3-, STAT5-, and AKT-dependent signaling pathways, which results in cellular proliferation and survival (Ma et al.). The majority of IL-2 is secreted by activated CD4+ and CD8+ T cells, although B cells and dendritic cells were found to produce IL-2 in small amounts. IL-2 downregulates immune responses to prevent autoimmunity during thymic development, influences the development of CD4+CD25+ regulatory T cells, and affects development of follicular helper T cells. IL-2 also controls inflammation by inhibiting Th17 differentiation (Banchereau et al.). Targeted deletions of the IL-2 gene in mice resulted in development of autoimmune hemolytic anemia, followed by ulcerative colitis. Similar effects were observed in mice that were deficient in IL-2 receptor α (Gaffen and Liu).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pakn:
    20 µG
  • Cat. No.:
    STMC78081
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    IL-2
  • Renhed:
    ≥95%
  • Species:
    Mouse

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.