Order Entry
Denmark
Orders LinkContactUsLinkComponent
Mouse Recombinant TNF-alpha

Mouse Recombinant TNF-alpha

100 µG
Katalog # STMC78069
Marketsource
Leverandør:   Stemcell Technologies
Mouse Recombinant TNF-alpha
Katalog # STMC78069
Leverandør:  Stemcell Technologies
Marketsource
CAS nummer:  
Mouse Recombinant TNF-alpha
Mouse Recombinant TNF-alpha

100 µG

CAS nummer:  
Price
kr6,632.00
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Tumor necrosis factor-α (TNF-α) is a pro-inflammatory cytokine that activates NF-κB, MAPK, and PI3K/AKT pathways. Activated T cells and macrophages are the primary producers of TNF-α in response to inflammation and infectious conditions. Many other cell types have been shown to produce TNF-α, among them B cells, NK cells, mast cells, neutrophils, dendritic cells, microglia, endothelial cells, smooth muscle cells, cardiomyocytes, and fibroblasts. TNF-α has cytotoxic effects on cancer cells in vitro by stimulating anti-tumor immunosuppressive responses. TNF-α stimulates expression of E- and P-selectins, thus facilitating adhesion of neutrophils, monocytes, and memory T cells to activated platelets and endothelial cells (Zelová andamp; Hosek). Other effects of TNF-α include vasodilatation and edema formation.

Advarsel: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifikationer

Specifikationer

  • Pakn:
    100 µG
  • Cat. No.:
    STMC78069
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    TNF-alpha
  • Renhed:
    ≥95%
  • Species:
    Mouse

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.