Orders LinkContactUsLinkComponent
SepMate™-15 (IVD)
SepMate™-15 (IVD)
SepMate™-15 (IVD), 500 units
Katalog # STMC85420
Marketsource
Leverandør:   Stemcell Technologies
SepMate™-15 (IVD)
Katalog # STMC85420
Leverandør:  Stemcell Technologies
Marketsource
SepMate™-15 (IVD)
SepMate™-15 (IVD)
SepMate™-15 (IVD), 500 units
Price
8.773,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
15 ml centrifuge tube with an insert for density gradient centrifugation
  • Eliminates the need for carefully layering blood over the density gradient medium (e.g. Lymphoprep™, and more)
  • Reduces total centrifuge time to 10 minutes with the brake on for fresh samples
  • Allows fast and easy harvesting of the isolated mononuclear cells by simply pouring off the supernatant
  • Can be combined with RosetteSep™ enrichment cocktails to isolate specific cell types in just 30 minutes
Simplify peripheral blood mononuclear cell to isolate specific immune cell subsets.
SepMate™ is manufactured under cGMP and registered as an In Vitro Diagnostic on SepMate™.
Advarsel: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifikationer
  • Beskrivelse:
    SepMate™-15 (IVD), 500 units
  • Seq:
    02