Orders LinkContactUsLinkComponent
[EN]STANDARD SOLUTION PH 4 140-4 1 * 4 L
[EN]STANDARD SOLUTION PH 4 140-4 1 * 4 L
Catalog # HORI3200174430
Supplier:   HORIBA
[EN]STANDARD SOLUTION PH 4 140-4 1 * 4 L
Catalog # HORI3200174430
Supplier:  HORIBA
[EN]STANDARD SOLUTION PH 4 140-4 1 * 4 L
Price
1.390,00 kr
Log in for availability
1 * 4 L