Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Carbon standards

Organic carbon standard, 250 ppm, 500 mL
Catalog # REGETOC250
Supplier:   REAGECON
Carbon standards
Catalog # REGETOC250
Supplier:  REAGECON
CAS Number:  
Carbon standards

Organic carbon standard, 250 ppm, 500 mL

CAS Number:  
Price
1.540,00 kr
Log in for availability
1 * 500 mL
Product Details & Documents

These standards are manufactured from high purity raw materials and is suitable for calibrating a wide range of Total Organic Carbon (TOC) and Total Inorganic Carbon (TIC) analysers to a high accuracy.

These standards are independent, traceable and certified.

Certifications: Accredited to IEC/ISO 17025 and ISO 17034.

Documents

Specifications

Specifications

  • Koncentration:
    250 ppm
  • Pakn:
    500 mL
  • Beskrivelse:
    Organic carbon standard

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.