Orders LinkContactUsLinkComponent
Accessories for GenePix® 4100A
Accessories for GenePix® 4100A
Red laser kit, 635 nm
Catalog # MLDV1-4040-0013
Supplier:   Molecular Devices
Accessories for GenePix® 4100A
Catalog # MLDV1-4040-0013
Supplier:  Molecular Devices
Accessories for GenePix® 4100A
Red laser kit, 635 nm
Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Beskrivelse:
    Red laser kit, 635 nm
  • Seq:
    02