BAM 7 ≥98%
25 mg
Katalog # CAYM14566-25
Leverandør: Cayman Chemical
Gå til:
Product Details & Documents
Synonymer: 3-Methyl-1-(4-phenyl-2-thiazolyl)-1H-pyrazole-4,5-dione 4-[2-(2-ethoxyphenyl)hydrazone], 4-(2-(2-Ethoxyphenyl)hydrazono)-5-methyl-2-(4-phenylthiazol-2-yl)-2,4-dihydro-3H-pyrazol-3-one, Bax Activator Molecule 7
- CAS nummer:331244-89-4
- Formel:C₂₁H₁₉N₅O₂S
- MDL nummer:MFCD01246556
- Molecular Weight:405.476 g/mol
- Opbevaringstemperatur:Fryser
Specifikationer
Specifikationer
- Pakn:25 mg



