Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
BAM 7 ≥98%

BAM 7 ≥98%

10 mg
Catalog # CAYM14566-10
Supplier:   Cayman Chemical
BAM 7 ≥98%
Catalog # CAYM14566-10
Supplier:  Cayman Chemical
CAS Number:  331244-89-4
BAM 7 ≥98%

10 mg

CAS Number:  331244-89-4
Price
1.860,00 kr
Log ind for tilgængelighed
1 * 10 mg
Product Details & Documents
Synonymer:  3-Methyl-1-(4-phenyl-2-thiazolyl)-1H-pyrazole-4,5-dione 4-[2-(2-ethoxyphenyl)hydrazone], 4-(2-(2-Ethoxyphenyl)hydrazono)-5-methyl-2-(4-phenylthiazol-2-yl)-2,4-dihydro-3H-pyrazol-3-one, Bax Activator Molecule 7
  • CAS nummer:331244-89-4
  • Formel:C₂₁H₁₉N₅O₂S
  • MDL nummer:MFCD01246556
  • Molecular Weight:405.476 g/mol
  • Opbevaringstemperatur:Fryser
Specifikationer

Specifikationer

  • Pakn:
    10 mg

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.