Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

BAM 7 ≥98%

1 mg
Catalog # CAYM14566-1
Supplier:   Cayman Chemical
BAM 7 ≥98%
Catalog # CAYM14566-1
Supplier:  Cayman Chemical
CAS Number:  331244-89-4
BAM 7 ≥98%

1 mg

CAS Number:  331244-89-4
Price
395,00 kr
Log in for availability
1 * 1 mg
Product Details & Documents
Synonyms:  3-Methyl-1-(4-phenyl-2-thiazolyl)-1H-pyrazole-4,5-dione 4-[2-(2-ethoxyphenyl)hydrazone], 4-(2-(2-Ethoxyphenyl)hydrazono)-5-methyl-2-(4-phenylthiazol-2-yl)-2,4-dihydro-3H-pyrazol-3-one, Bax Activator Molecule 7
  • CAS Number:331244-89-4
  • Formula:C₂₁H₁₉N₅O₂S
  • MDL Number:MFCD01246556
  • Molecular Weight:405.476 g/mol
  • Storage Temperature:Fryser
Specifications

Specifications

  • Pakn:
    1 mg

Specifications