Orders LinkContactUsLinkComponent
Yeast Extract Agar
Yeast Extract Agar
Yeast extract agar, 20 ml, Tube, 1
Katalog # LIOF34074
Leverandør:   LIOFILCHEM
Yeast Extract Agar
Katalog # LIOF34074
Leverandør:  LIOFILCHEM
CAS nummer:  
Yeast Extract Agar
Yeast extract agar, 20 ml, Tube, 1
CAS nummer:  
Price
423,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Medium for microorganisms enumeration in water.
Certifications: ISO6222 certified.
Specifikationer
Specifikationer
  • Pakning:
    Tube
  • Pakn:
    1
  • Beskrivelse:
    Yeast extract agar
  • Volumen:
    20 ml