Orders LinkContactUsLinkComponent
Accessories for CryoPlus Sample Storage Systems
Accessories for CryoPlus Sample Storage Systems
Rack for Matrix 2D plates measuring 58,42 mm tall, 699×135×93 mm, for CryoPlus 1, 2, 4, TSX40086FA, TSX50086FA, TSX60086FA, TSX70086FA
Katalog # BARN1950840
Leverandør:   Thermo Fisher Scientific
Accessories for CryoPlus Sample Storage Systems
Katalog # BARN1950840
Accessories for CryoPlus Sample Storage Systems
Rack for Matrix 2D plates measuring 58,42 mm tall, 699×135×93 mm, for CryoPlus 1, 2, 4, TSX40086FA, TSX50086FA, TSX60086FA, TSX70086FA
Price
1.270,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Rack for Matrix 2D plates measuring 58,42 mm tall, 699×135×93 mm, for CryoPlus 1, 2, 4, TSX40086FA, TSX50086FA, TSX60086FA, TSX70086FA
  • Seq:
    13